Tetracarbonyldi-µ-chlorodirhodium(I)

Supplier: STREM CHEMICALS INC MS
Warning

45-0450-2G 45-0450-500MG
100209-350EA 4161.23 USD
100209-350 100209-348
Tetracarbonyldi-µ-chlorodirhodium(I)
Tetracarbonyldi-µ-chlorodirhodium(I)
Formula: C₄Cl₂O₄Rh₂
Melting Pt: 124…125 °C
MDL Number: MFCD00135610
CAS Number: 14523-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR