Carbonylbis(triphenylphosphine)rhodium(I) chloride ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Carbonylchlorobis(triphenylphosphine)rhodium(I), Bis(triphenylphosphine)carbonylrhodium(I) Chloride, Chlorocarbonylbis(triphenylphosphine)rhodium(I)

45-0350-250MG 45-0350-5G 45-0350-1G
101442-334EA 120.31 USD
101442-334 100199-928 100206-674
Carbonylbis(triphenylphosphine)rhodium(I) chloride ≥99%
Carbonylbis(triphenylphosphine)rhodium(I) chloride
Formula: C₃₇H₃₀ClOP₂Rh
MW: 690.95 g/mol
Melting Pt: 170 °C (dec.)
MDL Number: MFCD00044273
CAS Number: 13938-94-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR