Bis[eta-(2,5-norbornadiene)]rhodium(I) tetrafluoroborate ≥96%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Bis(norbornadiene)rhodium(I) tetrafluoroborate, Bis(norbornadiene)rhodium (I) tetrafluoroborate

45-0230-250MG 45-0230-1G
100208-908EA 198.15 USD
100208-908 100208-910
Bis[eta-(2,5-norbornadiene)]rhodium(I) tetrafluoroborate ≥96%
Bis[eta-(2,5-norbornadiene)]rhodium(I) tetrafluoroborate
Formula: C₁₄H₈BF₄Rh
MW: 365.92 g/mol
MDL Number: MFCD00671775
CAS Number: 36620-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR