(R,R)-(-)-1,2-Bis[(o-methoxyphenyl)(phenyl)phosphino]ethane(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

45-0225-500MG 45-0225-100MG
100206-672EA 788.37 USD
100206-672 100206-670
(R,R)-(-)-1,2-Bis[(o-methoxyphenyl)(phenyl)phosphino]ethane(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate ≥95%
(R,R)-(-)-1,2-Bis[(o-methoxyphenyl)(phenyl)phosphino]ethane(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate
MDL Number: MFCD01074456
CAS Number: 56977-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR