1,2-Bis((2R,5R)-2,5-diisopropylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
45-0210-100MG
45-0210-2G
45-0210-500MG
101324-748EA
189.66
USD
101324-748
101324-752
101324-750
1,2-Bis((2R,5R)-2,5-diisopropylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate ≥98%
1,2-Bis((2R,5R)-2,5-diisopropylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate
BPE, DUPHOS
Formula:
C₃₄H₅₄BF₄P₂Rh |
MDL Number:
MFCD07369039 CAS Number: 569650-64-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...