(+)-1,2-Bis((2S,5S)-2,5-diphenylphospholano)ethane(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

45-0202-500MG 45-0202-2G 45-0202-100MG
101324-744EA 781.29 USD
101324-744 101324-746 101324-742
(+)-1,2-Bis((2S,5S)-2,5-diphenylphospholano)ethane(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate ≥98%
(+)-1,2-Bis((2S,5S)-2,5-diphenylphospholano)ethane(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate

BPE, DUPHOS

Formula: C₄₂H₄₈BF₄P₂Rh
MDL Number: MFCD07369038
CAS Number: 849950-53-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR