(-)-1,2-Bis((2R,5R)-2,5-diphenylphospholano)ethane(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

45-0201-100MG 45-0201-2G 45-0201-500MG
101324-736EA 181.17 USD
101324-736 101324-740 101324-738
(-)-1,2-Bis((2R,5R)-2,5-diphenylphospholano)ethane(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate ≥98%
(-)-1,2-Bis((2R,5R)-2,5-diphenylphospholano)ethane(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate

BPE, DUPHOS

Formula: C₄₂H₄₈BF₄P₂Rh
MW: 804.4882 g/mol
MDL Number: MFCD07369037
CAS Number: 528565-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR