3-Di-i-propylphosphoranylidene-2-(N,N-dimethylamino)-1H-indene(1,5-cyclooctadiene)rhodium(I) ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

45-0197-1G 45-0197-250MG
101419-036EA 986.52 USD
101419-036 101419-034
3-Di-i-propylphosphoranylidene-2-(N,N-dimethylamino)-1H-indene(1,5-cyclooctadiene)rhodium(I) ≥95%
3-Di-i-propylphosphoranylidene-2-(N,N-dimethylamino)-1H-indene(1,5-cyclooctadiene)rhodium(I)
MDL Number: MFCD09264278
CAS Number: 540492-55-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR