(Pentamethylcyclopentadienyl)rhodium(III) dichloride dimer ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

45-0195-2G 45-0195-500MG
100199-240EA 1840 USD
100199-240 100206-106
(Pentamethylcyclopentadienyl)rhodium(III) dichloride dimer ≥99%
(Pentamethylcyclopentadienyl)rhodium(III) dichloride dimer

CATHy Catalyst, Metallocenes, Derivatives & Cp Precursors

Formula: C₂₀H₃₀Cl₄Rh₂
MDL Number: MFCD00061552
CAS Number: 12354-85-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR