(2S,3S)-(+)-2,3-Bis(diphenylphosphino)bicyclo[2.2.1]hept-5-ene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (S,S)-NORPHOS-Rh

45-0185-100MG 45-0185-500MG
101433-220EA 188.25 USD
101433-220 101433-222
(2S,3S)-(+)-2,3-Bis(diphenylphosphino)bicyclo[2.2.1]hept-5-ene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate ≥97%
(2S,3S)-(+)-2,3-Bis(diphenylphosphino)bicyclo[2.2.1]hept-5-ene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate

NORPHOS

MDL Number: MFCD09839138
CAS Number: 78355-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR