(2R,3R)-(-)-2,3-Bis(diphenylphosphino)bicyclo[2.2.1]hept-5-ene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

45-0184-100MG 45-0184-500MG
101433-216EA 196.74 USD
101433-216 101433-218
(2R,3R)-(-)-2,3-Bis(diphenylphosphino)bicyclo[2.2.1]hept-5-ene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate ≥97%
(2R,3R)-(-)-2,3-Bis(diphenylphosphino)bicyclo[2.2.1]hept-5-ene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate

NORPHOS

MDL Number: MFCD09839137
CAS Number: 521272-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR