1,2-Bis((2S,5S)-2,5-dimethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
(1Z,5Z)-cycloocta-1,5-diene, (2S,5S)-1-[2-[(2S,5S)-2,5-dimethylphospholan-1-yl]ethyl]-2,5-dimethyl-phospholane, rhodium, tetrafluoroborate, (S,S)-ME-BPE-RH, (-)-1,2-Bis((2S,5S)-2,5-dimethylphospholano)ethane(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, ME-BPE-RH
45-0169-2G
45-0169-500MG
45-0169-100MG
100197-150EA
2516.55
USD
100197-150
100208-666
100208-664
1,2-Bis((2S,5S)-2,5-dimethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate ≥98%
1,2-Bis((2S,5S)-2,5-dimethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate
BPE, DUPHOS
Formula:
C₂₂H₃₈BF₄P₂Rh MW: 554.19 g/mol |
MDL Number:
MFCD09038435 CAS Number: 213343-65-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...