(+)-1,2-Bis((2R,5R)-2,5-dimethylphospholano)ethane(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 1,2-Bis((2R,5R)-2,5-dimethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate, (R,R)-ME-BPE-RH

45-0168-500MG 45-0168-100MG 45-0168-2G
100208-662EA 767.14 USD
100208-662 100208-660 100197-148
(+)-1,2-Bis((2R,5R)-2,5-dimethylphospholano)ethane(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate ≥98%
(+)-1,2-Bis((2R,5R)-2,5-dimethylphospholano)ethane(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate

BPE, DUPHOS

Formula: C₂₂H₄₀BF₄P₂Rh⁻
MW: 556.219 g/mol
MDL Number: MFCD09265087
CAS Number: 305818-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR