(+)-1,2-Bis((2S,5S)-2,5-dimethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

45-0159-2G 45-0159-500MG 45-0159-100MG
100197-142EA 2516.55 USD
100197-142 100208-650 100208-648
(+)-1,2-Bis((2S,5S)-2,5-dimethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate ≥98%
(+)-1,2-Bis((2S,5S)-2,5-dimethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate

BPE, DUPHOS

Formula: C₂₆H₃₈BF₄P₂Rh
MDL Number: MFCD01862467
CAS Number: 205064-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR