(-)-1,2-Bis((2R,5R)-2,5-dimethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
45-0158-100MG
45-0158-500MG
45-0158-2G
100208-644EA
181.17
USD
100208-644
100208-646
100197-140
(-)-1,2-Bis((2R,5R)-2,5-dimethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate ≥98%
(-)-1,2-Bis((2R,5R)-2,5-dimethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate
BPE, DUPHOS
Formula:
C₂₆H₃₈BF₄P₂Rh MW: 602.2376 g/mol |
MDL Number:
MFCD01862466 CAS Number: 210057-23-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...