(+)-1,2-Bis[(2S,5S)-2,5-diethylphospholano]benzene(cyclooctadiene)rhodium(I) trifluoromethanesulfonate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (+)-1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulphonate, (S,S)-Et-DUPHOS-Rh, 1,2-Bis[(2S,5S)-2,5-diethylphospholano]benzene(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulphonate

45-0151-100MG 45-0151-500MG 45-0151-2G
100208-640EA 189.66 USD
100208-640 100208-642 100197-138
(+)-1,2-Bis[(2S,5S)-2,5-diethylphospholano]benzene(cyclooctadiene)rhodium(I) trifluoromethanesulfonate ≥98%
(+)-1,2-Bis[(2S,5S)-2,5-diethylphospholano]benzene(cyclooctadiene)rhodium(I) trifluoromethanesulfonate

BPE, DUPHOS

Formula: C₃₁H₄₈F₃O₃P₂RhS
MW: 722.63 g/mol
MDL Number: MFCD00269861
CAS Number: 142184-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR