1,2-Bis((2R,5R)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

45-0148-2G 45-0148-100MG 45-0148-500MG
100197-132EA 2516.55 USD
100197-132 100208-628 100208-630
1,2-Bis((2R,5R)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate ≥98%
1,2-Bis((2R,5R)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate

BPE, DUPHOS

Formula: C₃₀H₄₈BF₄P₂Rh
MDL Number: MFCD01862464
CAS Number: 228121-39-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR