Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulphonate

45-0110-500MG 45-0110-2G
100203-164EA 502.46 USD
100203-164 100203-166
Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate ≥99%
Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate
Formula: C₁₇H₂₄F₃O₃RhS
Melting Pt: 159 °C (dec.)
MDL Number: MFCD00143752
CAS Number: 99326-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR