Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Bis(cycloocta-1,5-dien)rhodium(I)-tetrafluoroborat

45-0109-2G 45-0109-500MG
100206-666EA 1947.57 USD
100206-666 100198-800
Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate ≥98%
Bis(1,5-cyclooctadiene)rhodium (I) tetrafluoroborate
Formula: C₁₆H₂₄BF₄Rh
MW: 406.072 g/mol
MDL Number: MFCD00075045
CAS Number: 35138-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR