Acetylacetonatobis(cyclooctene)rhodium(I) ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

45-0016-500MG 45-0016-2G
100197-526EA 600.12 USD
100197-526 100207-400
Acetylacetonatobis(cyclooctene)rhodium(I) ≥97%
Acetylacetonatobis(cyclooctene)rhodium(I)
Formula: C₂₁H₃₆O₂Rh
MW: 434.4100 g/mol
MDL Number: MFCD06658137
CAS Number: 34767-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR