Acetylacetonato(1,5-cyclooctadiene)rhodium(I) ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
(Acetylacetonato)(1,5-cyclooctadiene)rhodium(I)
45-0010-100MG
45-0010-500MG
100196-236EA
92
USD
100196-236
100196-238
Acetylacetonato(1,5-cyclooctadiene)rhodium(I) ≥98%
Acetylacetonato(1,5-cyclooctadiene)rhodium(I)
Formula:
C₁₃H₂₀O₂Rh MW: 311.207 g/mol |
MDL Number:
MFCD00075046 CAS Number: 12245-39-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...