Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')ruthenium(III) ≥99% (99.9%-Ru)

Supplier: STREM CHEMICALS INC MS

Synonyms: Ru(TMHD)3, Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)ruthenium(III)

44-8000-5G 44-8000-1G
100198-798EA 1646.09 USD
100198-798 100198-796
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')ruthenium(III) ≥99% (99.9%-Ru)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')ruthenium(III)

Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD

Formula: C₃₃H₅₇O₆Ru
MW: 650.872 g/mol
Boiling Pt: 250 °C/ 120 °C (0,5 mmHg) (dec./ subl.)
Melting Pt: 210…213 °C
MDL Number: MFCD00269841
CAS Number: 38625-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR