Tris(acetonitrile)cyclopentadienylruthenium(II) hexafluorophosphate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: TRIS(ACETONITRILE)CYCLOPENTADIENYLRUTHENIUM(I) HEXACHLOROPHOSPHATE, Tris(acetonitrile)cyclopentadienylruthenium hexafluorophosphate, Trisacetonitrilecyclopentadienylrutheniumhexachlorophosphate

44-7870-250MG 44-7870-1G
100198-196EA 213.72 USD
100198-196 100204-060
Tris(acetonitrile)cyclopentadienylruthenium(II) hexafluorophosphate ≥98%
Tris(acetonitrile)cyclopentadienylruthenium(II) hexafluorophosphate

Metallocenes, Derivatives & Cp Precursors

Formula: C₁₁H₉F₆N₃PRu
MW: 429.24 g/mol
Melting Pt: 117…118 °C (dec.)
MDL Number: MFCD02684569
CAS Number: 80049-61-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR