Tricyclohexylphosphine[3-phenyl-1H-inden-1-ylidene][1,3-bis(2,4,6-trimethylphenyl)-4,5-dihydroimidazol-2-ylidene]ruthenium(II) dichloride ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: [1,3-Bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene]dichloro-(3-phenyl-1H-inden-1-ylidene)(tricyclohexylphosphine)ruthenium(II)

44-7777-100MG 44-7777-500MG
101419-022EA 101.91 USD
101419-022 101419-024
Tricyclohexylphosphine[3-phenyl-1H-inden-1-ylidene][1,3-bis(2,4,6-trimethylphenyl)-4,5-dihydroimidazol-2-ylidene]ruthenium(II) dichloride ≥95%
Tricyclohexylphosphine[3-phenyl-1H-inden-1-ylidene][1,3-bis(2,4,6-trimethylphenyl)-4,5-dihydroimidazol-2-ylidene]ruthenium(II) dichloride

Metathesis Catalyst, NHC

Formula: C₅₄H₆₉Cl₂N₂PRu
MW: 949.888 g/mol
MDL Number: MFCD08667892
CAS Number: 536724-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR