cis-Tetrakis(dimethylsulfoxide)dichlororuthenium(II) ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: cis-Dichlorotetrakis(dimethyl sulphoxide)ruthenium

44-7000-5G 44-7000-1G
100203-162EA 1167.69 USD
100203-162 100203-160
cis-Tetrakis(dimethylsulfoxide)dichlororuthenium(II) ≥98%
cis-Tetrakis(dimethylsulfoxide)dichlororuthenium(II)
Formula: C₈H₂₄Cl₂O₄RuS₄
MW: 484.49 g/mol
MDL Number: MFCD00050955
CAS Number: 59091-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR