Ruthenium 5% on carbon

Supplier: STREM CHEMICALS INC MS
Danger

44-4050-5G 44-4050-25G
100200-276EA 65.11 USD
100200-276 100196-880
Ruthenium 5% on carbon
Ruthenium
Formula: Ru
MW: 101.07
MDL Number: MFCD00011207
CAS Number: 7440-18-8
UN: 3178
ADR: 4.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR