Ruthenium 0,5% on carbon

Supplier: STREM CHEMICALS INC MS
Danger

44-4000-100G 44-4000-25G
100199-914EA 1058.71 USD
100199-914 100196-228
Ruthenium 0,5% on carbon
Ruthenium
Formula: Ru
MW: 101.07
MDL Number: MFCD00011207
CAS Number: 7440-18-8
UN: 3178
ADR: 4 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR