Ruthenium 5% on alumina

Supplier: STREM CHEMICALS INC MS
Warning

44-3910-25G 44-3910-100G
100201-656EA 297.23 USD
100201-656 100209-260
Ruthenium 5% on alumina
Ruthenium
Formula: Ru
MW: 101.07
MDL Number: MFCD00011207
CAS Number: 7440-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR