Ruthenium(III) nitrosylnitrate solution (1,5% Ru)

Supplier: STREM CHEMICALS INC MS
Warning

44-3800-250G 44-3800-50G 44-3800-1000G
100209-948EA 646.83 USD
100209-948 100209-950 100198-788
Ruthenium(III) nitrosylnitrate solution (1,5% Ru)
Ruthenium(III) nitrosylnitrate
Formula: N₄O₁₀Ru
MW: 317.09 g/mol
MDL Number: MFCD00016313
CAS Number: 34513-98-9
UN: 3218
ADR: 5.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR