Ruthenium(III) iodide, anhydrous ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

44-2500-1G 44-2500-5G
100211-092EA 145.78 USD
100211-092 100199-392
Ruthenium(III) iodide, anhydrous ≥98%
Ruthenium(III) iodide
Formula: I₃Ru
MW: 481.783 g/mol
MDL Number: MFCD00016316
CAS Number: 13896-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR