Dimethylammonium dichlorotri(mu-chloro)bis[(R)-(+)-2,2'-bis(diphenylphosphino)-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl]diruthenate(II)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: [NH2Me2][{RuCl((R)-H8-binap)}2(mu-Cl)3]

44-0516-50MG 44-0516-250MG
101418-992EA 117.48 USD
101418-992 101418-994
Dimethylammonium dichlorotri(mu-chloro)bis[(R)-(+)-2,2'-bis(diphenylphosphino)-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl]diruthenate(II)
Dimethylammonium dichlorotri(mu-chloro)bis[(R)-(+)-2,2'-bis(diphenylphosphino)-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl]diruthenate(II)

BINAP

Melting Pt: >100 °C (dec.)
MDL Number: MFCD09753036
CAS Number: 204933-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR