Dimethylammonium dichlorotri(mu-chloro)bis[(S)-(-)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]diruthenate(II)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: [NH2Me2][(RuCl((S)-tolbinap))2(mu-Cl)3], Dimethylammonium dichlorotri(mu-chloro)bis[(S)-(-)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]diruthenate(II), [NH2Me2][{RuCl((S)-tolbinap)}2(mu-Cl)3]

44-0513-1G 44-0513-250MG
101418-982EA 223.63 USD
101418-982 101418-980
Dimethylammonium dichlorotri(mu-chloro)bis[(S)-(-)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]diruthenate(II)
Dimethylammonium dichlorotri(mu-chloro)bis[(S)-(-)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]diruthenate(II)

BINAP

Formula: C₉₈H₈₈Cl₅NP₄Ru₂
MW: 1783.07 g/mol
Melting Pt: >100 °C (dec.)
MDL Number: MFCD09753034
CAS Number: 309735-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR