Dimethylammonium dichlorotri(mu-chloro)bis[(R)-(+)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]diruthenate(II)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
44-0512-250MG
44-0512-1G
101418-976EA
75.02
USD
101418-976
101418-978
Dimethylammonium dichlorotri(mu-chloro)bis[(R)-(+)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]diruthenate(II)
Dimethylammonium dichlorotri(mu-chloro)bis[(R)-(+)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]diruthenate(II)
BINAP
Formula:
C₉₈H₈₈Cl₅NP₄Ru₂ Melting Pt: >100 °C (dec.) |
MDL Number:
MFCD09753034 CAS Number: 749935-02-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...