Dimethylammonium dichlorotri(mu-chloro)bis[(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]diruthenate(II)

Supplier: STREM CHEMICALS INC MS
Warning

44-0511-1G 44-0511-250MG
101418-974EA 223.63 USD
101418-974 101418-972
Dimethylammonium dichlorotri(mu-chloro)bis[(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]diruthenate(II)
Dimethylammonium dichlorotri(mu-chloro)bis[(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]diruthenate(II)

BINAP

Formula: C₉₀H₇₂Cl₅NP₄Ru₂
Melting Pt: >100 °C (dec.)
MDL Number: MFCD09753033
CAS Number: 199541-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR