Dimethylammonium dichlorotri(mu-chloro)bis[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]diruthenate(II)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Dimethylammonium dichlorotri(mu-chloro)bis[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]diruthenate(II), [NH2Me2][{RuCl((R)-binap)}2(mu-Cl)3], [NH2Me2][(RuCl((R)-binap))2(mu-Cl)3]

44-0510-1G 44-0510-250MG
101418-970EA 223.63 USD
101418-970 101418-968
Dimethylammonium dichlorotri(mu-chloro)bis[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]diruthenate(II)
Dimethylammonium dichlorotri(mu-chloro)bis[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]diruthenate(II)

BINAP

Formula: C₉₀H₇₂Cl₅NP₄Ru₂
MW: 1670.86 g/mol
Melting Pt: >100 °C (dec.)
MDL Number: MFCD09753033
CAS Number: 199684-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR