Tricarbonyldichlororuthenium(II) dimer ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Dichlorotricarbonyl ruthenium(II) dimer, Dichlorotricarbonylruthenium(II) dimer

44-0450-100MG 44-0450-2G 44-0450-500MG
100205-128EA 107.57 USD
100205-128 100209-258 100208-904
Tricarbonyldichlororuthenium(II) dimer ≥98%
Tricarbonyldichlororuthenium(II) dimer
Formula: [RuCl₂(CO)₃]₂
Melting Pt: 208 °C (dec.)
MDL Number: MFCD00011528
CAS Number: 22594-69-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR