(+)-Dichloro[(4R)-4-(i-propyl)-2-{(R)-2-(diphenylphosphino)ferrocenyl}oxazoline](triphenylphosphine)ruthenium(II)

Supplier: STREM CHEMICALS INC MS

44-0442-100MG 44-0442-500MG
101418-960EA 140.12 USD
101418-960 101418-962
(+)-Dichloro[(4R)-4-(i-propyl)-2-{(R)-2-(diphenylphosphino)ferrocenyl}oxazoline](triphenylphosphine)ruthenium(II)
(+)-Dichloro[(4R)-4-(i-propyl)-2-{(R)-2-(diphenylphosphino)ferrocenyl}oxazoline](triphenylphosphine)ruthenium(II)
MDL Number: MFCD08459356
CAS Number: 1312582-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR