Dichloro(pentamethylcyclopentadienyl)ruthenium(III) polymer

Supplier: STREM CHEMICALS INC MS
Warning

44-0440-1G 44-0440-5G 44-0440-250MG
100197-128EA 167.02 USD
100197-128 100197-130 100209-946
Dichloro(pentamethylcyclopentadienyl)ruthenium(III) polymer
Dichloro(pentamethylcyclopentadienyl)ruthenium(III) polymer

Metallocenes, Derivatives & Cp Precursors

Formula: C₂₀H₃₀Cl₄Ru₂
MW: 614.4 g/mol
MDL Number: MFCD00070449
CAS Number: 96503-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR