Dichloro(1,5-cyclooctadiene)ruthenium(II), polymer ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
44-0425-1G
44-0425-5G
100206-646EA
151.45
USD
100206-646
100206-648
Dichloro(1,5-cyclooctadiene)ruthenium(II), polymer ≥97%
Dichloro(1,5-cyclooctadiene)ruthenium(II), polymer
Formula:
[RuCl₂(C₈H₁₂)]n Melting Pt: 200 °C (dec.) |
MDL Number:
MFCD00171304 CAS Number: 50982-12-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...