Dichloro[(S)-(-)-2,2',6,6'-tetramethoxy-4,4'-bis(di(3,5-xylyl)phosphino)-3,3'-bipyridine][(1S,2S)-(-)-1,2-diphenylethylenediamine]ruthenium(II) ≥95%

Supplier: STREM CHEMICALS INC MS

44-0391-500MG 44-0391-100MG
100198-786EA 631.26 USD
100198-786 100198-784
Dichloro[(S)-(-)-2,2',6,6'-tetramethoxy-4,4'-bis(di(3,5-xylyl)phosphino)-3,3'-bipyridine][(1S,2S)-(-)-1,2-diphenylethylenediamine]ruthenium(II) ≥95%
Dichloro[(S)-(-)-2,2',6,6'-tetramethoxy-4,4'-bis(di(3,5-xylyl)phosphino)-3,3'-bipyridine][(1S,2S)-(-)-1,2-diphenylethylenediamine]ruthenium(II)

P-PHOS

MDL Number: MFCD06658136
CAS Number: 821793-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR