(R)-Xyl-P-Phos RuCl2 (R,R)-DPEN ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Dichloro[(R)-(+)-2,2',6,6'-tetramethoxy-4,4'-bis(di(3,5-xylyl)phosphino)-3,3'-bipyridine][(1R,2R)-(+)-1,2-diphenylethylenediamine]ruthenium(II)

44-0390-100MG 44-0390-500MG
100198-780EA 164.18 USD
100198-780 100198-782
(R)-Xyl-P-Phos RuCl2 (R,R)-DPEN ≥95%
(R)-Xyl-P-Phos RuCl2 (R,R)-DPEN

P-PHOS

Formula: C₆₀H₆₆Cl₂N₄O₄P₂Ru
MW: 1141.13 g/mol
MDL Number: MFCD06658135
CAS Number: 478308-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR