Dichloro[(S)-(+)-4,12-bis(di(3,5-xylyl)phosphino)-[2.2]-paracyclophane][(1R,2R)-(+)-1,2-diphenylethylenediamine]ruthenium(II) ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

44-0381-10MG 44-0381-50MG
100209-944EA 147.2 USD
100209-944 100211-088
Dichloro[(S)-(+)-4,12-bis(di(3,5-xylyl)phosphino)-[2.2]-paracyclophane][(1R,2R)-(+)-1,2-diphenylethylenediamine]ruthenium(II) ≥95%
Dichloro[(S)-(+)-4,12-bis(di(3,5-xylyl)phosphino)-[2.2]-paracyclophane][(1R,2R)-(+)-1,2-diphenylethylenediamine]ruthenium(II)

PHANEPHOS

MDL Number: MFCD04974240
CAS Number: 364795-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR