Dichloro[(R)-(-)-4,12-bis(di(3,5-xylyl)phosphino)-[2.2]-paracyclophane][(1S,2S)-(-)-1,2-diphenylethylenediamine]ruthenium(II) ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

44-0380-50MG 44-0380-10MG
100211-084EA 291.57 USD
100211-084 100209-942
Dichloro[(R)-(-)-4,12-bis(di(3,5-xylyl)phosphino)-[2.2]-paracyclophane][(1S,2S)-(-)-1,2-diphenylethylenediamine]ruthenium(II) ≥95%
Dichloro[(R)-(-)-4,12-bis(di(3,5-xylyl)phosphino)-[2.2]-paracyclophane][(1S,2S)-(-)-1,2-diphenylethylenediamine]ruthenium(II)

PHANEPHOS

MDL Number: MFCD04974239
CAS Number: 325150-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR