Dichlorobis[2-(di-i-propylphosphino)ethylamine]ruthenium(II) ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

44-0265-1G 44-0265-250MG
101324-730EA 553.42 USD
101324-730 101324-728
Dichlorobis[2-(di-i-propylphosphino)ethylamine]ruthenium(II) ≥97%
Dichlorobis[2-(di-i-propylphosphino)ethylamine]ruthenium(II)
Formula: C₁₆H₄₀Cl₂N₂P₂Ru
MW: 494.4257 g/mol
MDL Number: MFCD07782005
CAS Number: 1092372-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR