Dichlorobis[2-(diphenylphosphino)ethylamine]ruthenium(II) ≥95% (mixture of isomers)

Supplier: STREM CHEMICALS INC MS

Synonyms: DIchlorobis(2-(diphenylphosphino)ethylamine)ruthenium(II)

44-0263-250MG 44-0263-1G
101324-724EA 144.37 USD
101324-724 101324-726
Dichlorobis[2-(diphenylphosphino)ethylamine]ruthenium(II) ≥95% (mixture of isomers)
Dichlorobis[2-(diphenylphosphino)ethylamine]ruthenium(II)
Formula: C₂₈H₃₂Cl₂N₂P₂Ru
MW: 630.4906 g/mol
MDL Number: MFCD07782004
CAS Number: 506417-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR