Dichlorobis(2-(di-tert-butylphosphino)ethylamine)ruthenium(II) ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Dichlorobis[2-(di-t-butylphosphino)ethylamine]ruthenium(II), DICHLOROBIS[2-(DI-T-BUTYLPHOSPHINO)ETHYLAMINE]RUTHENIUM(II)

44-0260-250MG 44-0260-1G
101324-720EA 67.94 USD
101324-720 101324-722
Dichlorobis(2-(di-tert-butylphosphino)ethylamine)ruthenium(II) ≥97%
Dichlorobis(2-(di-tert-butylphosphino)ethylamine)ruthenium(II)
Formula: C₂₀H₄₈Cl₂N₂P₂Ru
MW: 550.536 g/mol
MDL Number: MFCD07782003
CAS Number: 1092372-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR