[(R)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)dichloride ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Dichloro[(R)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II), Dichloro[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II), Dichloro [(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphathyl] ruthenium(II)

44-0248-500MG 44-0248-2G
100208-900EA 294.4 USD
100208-900 100208-902
[(R)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)dichloride ≥95%
[(R)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)dichloride

BINAP

Formula: C₄₄H₃₂Cl₂P₂Ru
MW: 794.66 g/mol
MDL Number: MFCD01073794
CAS Number: 132071-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR