Dichloro[(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl][(1R,2R)-(+)-1,2-diphenylethylenediamine]ruthenium(II) ≥98%

Supplier: STREM CHEMICALS INC MS

44-0222-250MG 44-0222-10MG 44-0222-50MG
100199-904EA 1677.23 USD
100199-904 100206-642 100199-902
Dichloro[(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl][(1R,2R)-(+)-1,2-diphenylethylenediamine]ruthenium(II) ≥98%
Dichloro[(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl][(1R,2R)-(+)-1,2-diphenylethylenediamine]ruthenium(II)

BINAP

Melting Pt: >100 °C (dec.)
MDL Number: MFCD02684565
CAS Number: 329735-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR