Dichloro[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl][(1R,2R)-(+)-1,2-diphenylethylenediamine]ruthenium(II) ≥90%

Supplier: STREM CHEMICALS INC MS

44-0220-10MG 44-0220-50MG 44-0220-250MG
100206-638EA 208.06 USD
100206-638 100199-894 100199-896
Dichloro[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl][(1R,2R)-(+)-1,2-diphenylethylenediamine]ruthenium(II) ≥90%
Dichloro[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl][(1R,2R)-(+)-1,2-diphenylethylenediamine]ruthenium(II)

BINAP

Melting Pt: >100 °C (dec.)
MDL Number: MFCD02684565
CAS Number: 212143-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR