Dichloro[(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl][(2S)-(+)-1,1-bis(4-methoxyphenyl)-3-methyl-1,2-butanediamine]ruthenium(II) dichloromethane adduct ≥97%

Supplier: STREM CHEMICALS INC MS

44-0211-10MG 44-0211-250MG 44-0211-50MG
100206-636EA 242.03 USD
100206-636 100199-892 100199-890
Dichloro[(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl][(2S)-(+)-1,1-bis(4-methoxyphenyl)-3-methyl-1,2-butanediamine]ruthenium(II) dichloromethane adduct ≥97%
Dichloro[(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl][(2S)-(+)-1,1-bis(4-methoxyphenyl)-3-methyl-1,2-butanediamine]ruthenium(II) dichloromethane adduct

BINAP

Melting Pt: >100 °C (dec.)
MDL Number: MFCD02684564
CAS Number: 212143-24-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR